Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX23 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982030
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2030 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2030 Supplier Bioss Supplier No. BS12199R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PC4 (activated RNA polymerase II transcription cofactor 4) is a transcriptional coactivator, possessing the ability to suppress promoter-driven as well as nonspecific transcription via its DNA binding activity. The repressive activity of PC4 on promoter-driven transcription is alleviated by transcription factor TFIIH. TFIIH protects promoters from PC4-mediated repression by relieving the topological constraint imposed by PC4 through the ERCC3 helicase activity.

Specifications

Antigen DDX23
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Ddx23
Gene Accession No. Q9BUQ8
Gene Alias 100 kDa U5 snRNP-specific protein; 3110082M05Rik; 4921506D17Rik; DDX23; DEAD (Asp-Glu-Ala-Asp) box polypeptide 23; DEAD box protein 23; DEAD-box helicase 23; Probable ATP-dependent RNA helicase DDX23; prp28; PRP28 homolog; PRP28 homolog, yeast; PRP28p homolog; PRPF28; SNRNP100; U5 snRNP 100 kD protein; U5-100K; U5-100kD
Gene Symbols Ddx23
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DDX23.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9416
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.