Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX24 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002225AF405
Description
DDX24 Polyclonal antibody specifically detects DDX24 in Human samples. It is validated for Western BlotSpecifications
DDX24 | |
Polyclonal | |
Western Blot | |
ATP-dependent RNA helicase DDX24, DEAD (Asp-Glu-Ala-Asp) box polypeptide 24, DEAD box protein 24, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 24, EC 3.6.1, EC 3.6.4.13, S. cerevisiae CHL1-like helicase | |
The immunogen recognized by this antibody maps to a region between residue 325 and 375 of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 24 using the numbering given in entry NP_065147.1 (GeneID 57062). | |
0.1 mL | |
Cell Cycle and Replication | |
57062 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction