Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX24 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002225PE
Description
DDX24 Polyclonal antibody specifically detects DDX24 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| DDX24 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| ATP-dependent RNA helicase DDX24, DEAD (Asp-Glu-Ala-Asp) box polypeptide 24, DEAD box protein 24, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 24, EC 3.6.1, EC 3.6.4.13, S. cerevisiae CHL1-like helicase | |
| Maps to a region between residue 325 and 375 of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 24 using the numbering given in entry NP_065147.1 (GeneID 57062). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 57062 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction