Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541286
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541286 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541286 Supplier Invitrogen™ Supplier No. PA541286
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ELRHLLSQPL FTESQKTKYP TQSGKPPLLV SAPSKSESAL SCLSKQKKKK Sequence homology: Human: 100%; Mouse: 79%; Rat: 79%.

METAP1 removes the amino-terminal methionine from nascent proteins. METAP1 is required for normal progression through the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX24
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Ddx24
Gene Accession No. Q9GZR7
Gene Alias 1700055J08Rik; 2510027P10Rik; AI649272; ATP-dependent RNA helicase DDX24; DDX24; DEAD (Asp-Glu-Ala-Asp) box helicase 24; DEAD (Asp-Glu-Ala-Asp) box polypeptide 24; DEAD box protein 24; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 13 (RNA helicase A); DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 24; DEAD-box helicase 24; LOC553462 protein; S. cerevisiae CHL1-like helicase; si:ch1073-416d2.2
Gene Symbols Ddx24
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human DDX24.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57062
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.