Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551721
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551721 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551721 Supplier Invitrogen™ Supplier No. PA551721
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FPVQTKYMDV VKERIRLARQ IEKSEYRNFQ ACLHNSWIEQ AAAALEIELE EDMYKGGKAD QQEERRRQKQ MKVLKKELRH LLSQPLFTES QKTKYPTQSG KPPLLVSAPS KSESALSCLS KQKKKKTKKP KE Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.

METAP1 removes the amino-terminal methionine from nascent proteins. METAP1 is required for normal progression through the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX24
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ddx24
Gene Accession No. Q9GZR7
Gene Alias 1700055J08Rik; 2510027P10Rik; AI649272; ATP-dependent RNA helicase DDX24; DDX24; DEAD (Asp-Glu-Ala-Asp) box helicase 24; DEAD (Asp-Glu-Ala-Asp) box polypeptide 24; DEAD box protein 24; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 13 (RNA helicase A); DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 24; DEAD-box helicase 24; LOC553462 protein; S. cerevisiae CHL1-like helicase; si:ch1073-416d2.2
Gene Symbols Ddx24
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DDX24. Recombinant protein control fragment (Product #RP-89021).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57062
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.