Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120368
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120368 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120368 Supplier Thermo Scientific Supplier No. PA5120368
Add to Cart
Add to Cart
This item is not returnable. View return policy

DDX24 Polyclonal Antibody

DDX24 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX24
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Ddx24
Gene Accession No. Q9ESV0, Q9GZR7
Gene Alias 1700055J08Rik; 2510027P10Rik; AI649272; ATP-dependent RNA helicase DDX24; DDX24; DEAD (Asp-Glu-Ala-Asp) box helicase 24; DEAD (Asp-Glu-Ala-Asp) box polypeptide 24; DEAD box protein 24; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 13 (RNA helicase A); DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 24; DEAD-box helicase 24; LOC553462 protein; S. cerevisiae CHL1-like helicase; si:ch1073-416d2.2
Gene Symbols Ddx24
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 510-859 of human DDX24 (NP_065147.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27225, 373065, 57062
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.