Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554159
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554159 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554159 Supplier Invitrogen™ Supplier No. PA554159
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NSHFSNLSQP RKNLWGIKST AVRNIDGSIN NINEDDEEDV VDLAANSLLN KLIHQSLVES SHRVEVLQKD PSSP Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 82%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX25
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DDX25
Gene Accession No. Q9UHL0
Gene Alias ATP-dependent RNA helicase DDX25; AW047046; Ddx25; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 25; DEAD (Asp-Glu-Ala-Asp) box helicase 25; DEAD (Asp-Glu-Ala-Asp) box polypeptide 25; DEAD box protein 25; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 25; DEAD-box helicase 25; gonadotropin-regulated testicular RNA helicase; GRTH
Gene Symbols DDX25
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DDX25. Recombinant protein control fragment (Product #RP-91724).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29118
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.