Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX26B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553044
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553044 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553044 Supplier Invitrogen™ Supplier No. PA553044
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DNLDCGLSYS VISYLKKLSQ QTKLESERIL ASVGKKPPQE IGIKVKNHSG GGMSLTHNKN FRKLLKEITG ETALRLTELN TKEFAGFQIG LLNKDLKPQT YRNAYDIPRR GLLDQLTRMR SNLLKTH Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 76%.

DDX26B is a protein coding gene. An important paralog of this gene is INTS6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX26B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene INTS6L
Gene Accession No. Q5JSJ4
Gene Alias 4930535D10Rik; 6330505F04Rik; D130066O12; ddx26b; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 26B; hypothetical protein MGC63527; int6; Integrator complex subunit 6; integrator complex subunit 6 like; integrator complex subunit 6-like; ints6; Ints6l; LOW QUALITY PROTEIN: integrator complex subunit 6-like; protein DDX26B; zgc:63527
Gene Symbols INTS6L
Host Species Rabbit
Immunogen Recombinant protein fragment of Human INTS6L (Product #RP-92227)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203522
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.