Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX26B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541305
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541305 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541305 Supplier Invitrogen™ Supplier No. PA541305
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ASTEPEQLGS VPTDESAITQ MCEVTGGRSY CVRTQRMLNQ CLESLVQKVQ Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

DDX26B is a protein coding gene. An important paralog of this gene is INTS6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX26B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene INTS6L
Gene Accession No. Q5JSJ4
Gene Alias 4930535D10Rik; 6330505F04Rik; D130066O12; ddx26b; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 26B; hypothetical protein MGC63527; int6; Integrator complex subunit 6; integrator complex subunit 6 like; integrator complex subunit 6-like; ints6; Ints6l; LOW QUALITY PROTEIN: integrator complex subunit 6-like; protein DDX26B; zgc:63527
Gene Symbols INTS6L
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human DDX26B (aa 179-228).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203522
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.