Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX27 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP126601AF532
Description
DDX27 Polyclonal antibody specifically detects DDX27 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
DDX27 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
DEAD (Asp-Glu-Ala-Asp) box polypeptide 27, DEAD box protein 27, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 27, dJ686N3.1, DKFZp667N057, DRS1, EC 3.6.1, EC 3.6.4.13, FLJ12917, FLJ20596, FLJ22238, MGC1018, MGC163147, PP3241, probable ATP-dependent RNA helicase DDX27, RHLP, RNA helicase-like protein, Rrp3p | |
The immunogen recognized by this antibody maps to a region between residue 700 and 750 of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 27 using the numbering given in entry NP_060365.6 (GeneID 55661). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
55661 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction