Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX27 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP126601
Description
DDX27 Polyclonal specifically detects DDX27 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative).Specifications
DDX27 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
DEAD (Asp-Glu-Ala-Asp) box polypeptide 27, DEAD box protein 27, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 27, dJ686N3.1, DKFZp667N057, DRS1, EC 3.6.1, EC 3.6.4.13, FLJ12917, FLJ20596, FLJ22238, MGC1018, MGC163147, PP3241, probable ATP-dependent RNA helicase DDX27, RHLP, RNA helicase-like protein, Rrp3p | |
Rabbit | |
Affinity Purified | |
RUO | |
55661 | |
Human, Mouse | |
IgG |
Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
1.0 mg/mL | |
Immunohistochemistry 1:1000 - 1:5000, Immunoprecipitation 5-10 ug/mg lysate, Immunohistochemistry-Paraffin 1:1000 - 1:5000, Western Blot (Negative) | |
NP_060365.6 | |
DDX27 | |
The immunogen recognized by this antibody maps to a region between residue 700 and 750 of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 27 using the numbering given in entry NP_060365.6 (GeneID 55661). | |
100 μL | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, and Northern white-cheeked gibbon. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction