Learn More
DDX27 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc IHC00634T

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Monkey, Gorilla, Chimpanzee, Northern white-cheeked gibbon, Crab-eating macaque and Wh
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. DDX27 is a DEAD box protein, the function of which has not been determined.Specifications
DDX27 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q921N6, Q96GQ7 | |
DDX27 | |
Between 700 and 750 | |
10 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
DDX27 | |
DEAD (Asp-Glu-Ala-Asp) box polypeptide 27, DRS1, RHLP, Rrp3p, PP3241, HSPC259, dJ686N3.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
228889, 55661 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.