Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX27 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $485.00
Specifications
Antigen | DDX27 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
DDX27 Polyclonal specifically detects DDX27 in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DDX27 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
55661 | |
IgG | |
Affinity purified |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Rat | |
DEAD (Asp-Glu-Ala-Asp) box polypeptide 27, DEAD box protein 27, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 27, dJ686N3.1, DKFZp667N057, DRS1, EC 3.6.1, EC 3.6.4.13, FLJ12917, FLJ20596, FLJ22238, MGC1018, MGC163147, PP3241, probable ATP-dependent RNA helicase DDX27, RHLP, RNA helicase-like protein, Rrp3p | |
Recombinant fusion protein containing a sequence corresponding to amino acids 587-796 of human DDX27 (NP_060365.7). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title