Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX28 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002345AF405
Description
DDX28 Polyclonal antibody specifically detects DDX28 in Human samples. It is validated for Western BlotSpecifications
DDX28 | |
Polyclonal | |
Western Blot | |
DEAD (Asp-Glu-Ala-Asp) box polypeptide 28, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 28, EC 3.6.1, EC 3.6.4.13, FLJ11282, M, MDDX28probable ATP-dependent RNA helicase DDX28, Mitochondrial DEAD box protein 28, mitochondrial DEAD-box polypeptide 28 | |
The immunogen recognized by this antibody maps to a region between residue 500 and the C-terminus (residue 540) of human DEAD (Asp-Glu-Ala-Asp) Box Polypeptide 28 using the numbering given in entry NP_060850.1 (GeneID 55794). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
55794 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction