Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX28 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530762
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530762 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530762 Supplier Invitrogen™ Supplier No. PA530762
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32. Predicted reactivity: Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

WDR11 is a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene is located in the chromosome 10q25-26 region, which is frequently deleted in gliomas and tumors of other tissues, and is disrupted by the t(10;19) translocation rearrangement in glioblastoma cells. The gene location suggests that it is a candidate gene for the tumor suppressor locus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX28
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene DDX28
Gene Accession No. Q9NUL7
Gene Alias 2410004K13Rik; AI449652; ATP-dependent RNA helicase Ddx28; DDX28; DEAD (Asp-Glu-Ala-Asp) box polypeptide 28; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 28; DEAD-box helicase 28; MDDX28; Mitochondrial DEAD box protein 28; mitochondrial DEAD-box polypeptide 28; probable ATP-dependent RNA helicase DDX28
Gene Symbols DDX28
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human DDX28. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55794
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.