Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX28 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584163
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584163 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584163 Supplier Invitrogen™ Supplier No. PA584163
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VAELVHILKH RDRAERTGPS GTVLVFCNSS STVNWLGYIL DDHKIQHLRL QGQMPALMRV GIFQSFQKSS RDILLCTDIA SRGLDSTGV Highest antigen sequence indentity to the following orthologs: Mouse - 81%, Rat - 79%.

WDR11 is a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene is located in the chromosome 10q25-26 region, which is frequently deleted in gliomas and tumors of other tissues, and is disrupted by the t(10;19) translocation rearrangement in glioblastoma cells. The gene location suggests that it is a candidate gene for the tumor suppressor locus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX28
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DDX28
Gene Accession No. Q9NUL7
Gene Alias 2410004K13Rik; AI449652; ATP-dependent RNA helicase Ddx28; DDX28; DEAD (Asp-Glu-Ala-Asp) box polypeptide 28; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 28; DEAD-box helicase 28; MDDX28; Mitochondrial DEAD box protein 28; mitochondrial DEAD-box polypeptide 28; probable ATP-dependent RNA helicase DDX28
Gene Symbols DDX28
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DDX28. Recombinant protein control fragment (Product #RP-106478).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55794
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.