Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX3 Antibody (11F11D10), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP214848PE
Description
DDX3 Monoclonal specifically detects DDX3 in Human samples. It is validated for Flow Cytometry.Specifications
DDX3 | |
Monoclonal | |
PE | |
O00571 | |
DDX3X | |
A partial recombinant protein containing amino acids 24-203 of human DDX3 was used as immunogen for this antibody. | |
0.1 mL | |
Cancer, Tumor Suppressors | |
1654 | |
Store at 4C in the dark. | |
IgG2b κ |
Flow Cytometry | |
11F11D10 | |
Flow Cytometry | |
DBXCAP-Rf, DDX14, DDX3DEAD/H box-3, DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, X-linked, DEAD box protein 3, X-chromosomal, DEAD box, X isoform, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 3, EC 3.6.1, EC 3.6.4.13, helicase like protein 2, Helicase-like protein 2, HLP2ATP-dependent RNA helicase DDX3X | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction