Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX3 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB200196R
Description
DDX3 Polyclonal antibody specifically detects DDX3 in Human samples. It is validated for Western BlotSpecifications
DDX3 | |
Polyclonal | |
Western Blot | |
DBXCAP-Rf, DDX14, DDX3DEAD/H box-3, DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, X-linked, DEAD box protein 3, X-chromosomal, DEAD box, X isoform, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 3, EC 3.6.1, EC 3.6.4.13, helicase like protein 2, Helicase-like protein 2, HLP2ATP-dependent RNA helicase DDX3X | |
The immunogen recognized by this antibody maps to a region between residue 600 and the C-terminus (residue 661) of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 3 using the numbering given in entry NP_001347.2 (GeneID 1654). | |
0.1 mL | |
Cancer, Tumor Suppressors | |
1654 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction