Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX31 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541291
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541291 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541291 Supplier Invitrogen™ Supplier No. PA541291
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QASSEAPPAK RRNETSFLPA KKTSVKETQR TFKGNAQKMF SPKKHSVSTS Sequence homology: Guinea Pig: 83%; Human: 100%; Mouse: 86%; Rat: 93%.

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. DDX31 is a member of this family. The function of this member has not been determined. Alternative splicing of this gene generates 2 transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX31
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Ddx31
Gene Accession No. Q9H8H2
Gene Alias 5830444G11Rik; DDX31; DEAD (Asp-Glu-Ala-Asp) box polypeptide 31; DEAD box protein 31; DEAD/DEXH helicase DDX31; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 31; DEAD-box helicase 31; G2 helicase; Gm997; Helicain; PPP1R25; Probable ATP-dependent RNA helicase DDX31; protein phosphatase 1, regulatory subunit 25
Gene Symbols Ddx31
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human DDX31 (aa 140-189).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64794
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.