Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX39 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576309
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576309 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576309 Supplier Thermo Scientific Supplier No. PA576309
Add to Cart
Add to Cart

DDX39 Polyclonal Antibody

DDX39 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX39
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene DDX39A
Gene Accession No. O00148, Q5U216, Q8VDW0
Gene Alias 2610307C23Rik; ATP-dependent RNA helicase DDX39; ATP-dependent RNA helicase DDX39A; BAT1; BAT1L; DDX39; Ddx39a; Ddxl; DEAD (Asp-Glu-Ala-Asp) box polypeptide 39; DEAD (Asp-Glu-Ala-Asp) box polypeptide 39A; DEAD box protein 39; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 39; DEAD-box helicase 39A; nuclear RNA helicase URH49; Nuclear RNA helicase, DECD variant of DEAD box family; UAP56-related helicase, 49 kDa; URH49
Gene Symbols Ddx39, DDX39A
Host Species Rabbit
Immunogen DDX39A fusion protein.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10212, 68278, 89827
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.