Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DDX39A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109624
Description
Immunogen sequence: MAEQDVENDL LDYDEEEEPQ APQESTPAPP KKDIKGSYVS IHSSGFRDFL LKPELLRAIV DCGFEHPSEV QHECIPQAIL GMDVLCQAKS GMGKTAVFVL.
This gene encodes a member of the DEAD box protein family. These proteins are characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD) and are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of the DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene is thought to play a role in the prognosis of patients with gastrointestinal stromal tumors. A pseudogene of this gene is present on chromosome 13. Alternate splicing results in multiple transcript variants. Additional alternatively spliced transcript variants of this gene have been described, but their full-length nature is not known.
Specifications
DDX39A | |
Polyclonal | |
Unconjugated | |
DDX39A | |
2610307C23Rik; ATP-dependent RNA helicase DDX39; ATP-dependent RNA helicase DDX39A; BAT1; BAT1L; DDX39; Ddx39a; Ddxl; DEAD (Asp-Glu-Ala-Asp) box polypeptide 39; DEAD (Asp-Glu-Ala-Asp) box polypeptide 39A; DEAD box protein 39; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 39; DEAD-box helicase 39A; nuclear RNA helicase URH49; Nuclear RNA helicase, DECD variant of DEAD box family; UAP56-related helicase, 49 kDa; URH49 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10212, 68278, 89827 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.81 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00148, Q5U216, Q8VDW0 | |
Ddx39, DDX39A | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human DDX39A (NP_0057952). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction