Learn More
DDX39B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 147981AP150UL

Description
This antibody may cross-react with DDX39A.
This gene encodes a member of the DEAD box family of RNA-dependent ATPases that mediate ATP hydrolysis during pre-mRNA splicing. The encoded protein is an essential splicing factor required for association of U2 small nuclear ribonucleoprotein with pre-mRNA, and also plays an important role in mRNA export from the nucleus to the cytoplasm. A cluster of genes, BAT1-BAT5, is localized in the vicinity of the genes for TNF alpha and TNF beta. These genes are all within the human major histocompatibility complex class III region. Mutations in this gene may be associated with rheumatoid arthritis. Alternatively spliced transcript variants encoding the same protein have been described.Specifications
DDX39B | |
Polyclonal | |
Unconjugated | |
DDX39B | |
ATP dependent RNA helicase p47, BAT1, D6S81E, DDX39B, DEAD box protein UAP56, HLA B associated transcript 1, Spliceosome RNA helicase BAT1, UAP56 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114612, 53817, 7919 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13838, Q63413, Q9Z1N5 | |
Ddx39b | |
DDX39B Fusion Protein Ag6512 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.