Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX3Y Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590055
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590055 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590055 Supplier Invitrogen™ Supplier No. PA590055
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSHVVVKNDP ELDQQLANLD LNSEKQSGGA STASKGRYIP PHLRNREASK GFHDKDSSGW SCSKDKDAYS SFGSRDSRGK PGYFSERGSG SRGRFDDRGR SDYDGIGNRE RPGFGRFERS GHSRWCDKSV EDDWSKPLPP SERLEQELFS GGNTGINFEK YDDIPVEATG SNCPPHIENF SDIDMGEIIM GNIELTRYTR PTPVQKHAIP; Positive Samples: A-549, HeLa, rat testis; Cellular Location: Cytoplasm, Nucleus.

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases which are implicated in a number of cellular processes involving RNA binding and alteration of RNA secondary structure. DDX3Y is one member of DEAD box RNA helicases, encode by the gene locate on the chromosome Y in the azoospermia factor A region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX3Y
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.50 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Ddx3y
Gene Accession No. O15523
Gene Alias 8030469F12Rik; ATP-dependent RNA helicase DDX3Y; D1Pas1 related sequence 1; D1Pas1-related sequence 1; D1Pas1-rs1; DBY; Ddx3y; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide, Y chromosome; DEAD (Asp-Glu-Ala-Asp) box helicase 3, Y-linked; DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, Y-linked; DEAD box protein 3, Y-chromosomal; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, Y chromosome; Dead2; DEAD-box helicase 3, Y-linked; DEAD-box RNA helicase DEAD2; mDEAD2; RGD1309586; uncharacterized protein LOC364073
Gene Symbols Ddx3y
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human DDX3Y (NP_0011161371).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 364073, 8653
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.