Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DDX3Y Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140228
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: NDPELDQQLA NLDLNSEKQS GGASTA.
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases which are implicated in a number of cellular processes involving RNA binding and alteration of RNA secondary structure. DDX3Y is one member of DEAD box RNA helicases, encode by the gene locate on the chromosome Y in the azoospermia factor A region.
Specifications
DDX3Y | |
Polyclonal | |
Unconjugated | |
Ddx3y | |
8030469F12Rik; ATP-dependent RNA helicase DDX3Y; D1Pas1 related sequence 1; D1Pas1-related sequence 1; D1Pas1-rs1; DBY; Ddx3y; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide, Y chromosome; DEAD (Asp-Glu-Ala-Asp) box helicase 3, Y-linked; DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, Y-linked; DEAD box protein 3, Y-chromosomal; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, Y chromosome; Dead2; DEAD-box helicase 3, Y-linked; DEAD-box RNA helicase DEAD2; mDEAD2; RGD1309586; uncharacterized protein LOC364073 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8653 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15523 | |
Ddx3y | |
Recombinant protein corresponding to Human DDX3Y. Recombinant protein control fragment (Product #RP-108972). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction