Learn More
Invitrogen™ DDX4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579147
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat testis tissue, mouse testis tissue. IHC: Rat Ovary tissue, Rat Testis tissue, human ovarian cancer tissue IHC-F: Rat Intestine Tissue. ICC/IF: U20S cell, MCF-7 Cell. Flow: PC-3 cell, SiHa cell.
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp, are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is a homolog of VASA proteins in Drosophila and several other species. The gene is specifically expressed in the germ cell lineage in both sexes and functions in germ cell development. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
DDX4 | |
Polyclonal | |
Unconjugated | |
DDX4 | |
ATP-dependent RNA helicase DDX4; AV206478; Cvh; Ddx4; D-E-A-D (aspartate-glutamate-alanine-aspartate) box polypeptide 4; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 4; DEAD (Asp-Glu-Ala-Asp) box polypeptide 4; DEAD box polypeptide 4; DEAD box protein 4; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 4; DEAD-box helicase 4; MGC111074; Mvh; mvh / m'vasa; probable ATP-dependent RNA helicase DDX4; putative ATP-dependent RNA helicase DDX4; RVLG; VASA; Vasa homolog; vasa-like gene protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13206, 310090, 54514 | |
-20°C | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
Q61496, Q64060, Q9NQI0 | |
DDX4 | |
A synthetic peptide corresponding to a sequence in the middle region of human DDX4 (253-272aa EDEDSIFAHYQTGINFDKYD). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.