Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX41 Antibody (PCRP-DDX41-1B4), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308520F
Description
DDX41 Monoclonal antibody specifically detects DDX41 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein ArraySpecifications
DDX41 | |
Monoclonal | |
FITC | |
ABSDEAD-box protein abstrakt, DEAD (Asp-Glu-Ala-Asp) box polypeptide 41, DEAD box protein 41, DEAD box protein abstrakt homolog, EC 3.6.1, EC 3.6.4.13,2900024F02Rik, MGC8828, probable ATP-dependent RNA helicase DDX41, putative RNA helicase | |
Recombinant full-length human DDX41 protein (Uniprot: Q9UJV9) | |
100 μg | |
Apoptosis | |
51428 | |
Store at 4°C in the dark. | |
IgG2b |
Western Blot, Flow Cytometry, Immunofluorescence, Protein Array | |
PCRP-DDX41-1B4 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction