Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ DDX41, Mouse, Unlabeled, Clone: 4D8.2,

Catalog No. MABF1107MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABF1107MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF1107MI Supplier MilliporeSigma™ Supplier No. MABF1107

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Probable ATP-dependent RNA helicase DDX41 (UniProt: Q9UJV9; also known as DEAD box protein 41; DEAD box protein abstrakt homolog) is encoded by DDX41 (also known as ABS) gene (Gene ID 51428) in human. DDX41 belongs to the family of DEAD box helicase family of proteins, characterized by the presence of the conserved Asp-Glu-Ala-Asp (DEAD) motif. DDX41 is reported to sense DNA virus in myeloid dendritic cells (mDCs) mediated by the adaptor STING (stimulator of interferon genes). DDX41 is reported to bind to both DNA and STING through a DEADc domain and then DDX41-STING complex localizes to the cytoplasm. DDX41 binds bacterial DNA in the infected cells and is shown to be more important than gamma-interferon-inducible protein IFI16 in the initial sensing of cytosolic B-form DNA in triggering the early burst of type I interferon response. Knockdown of DDX41 expression blocks the ability of mDCs to mount type I interferon and cytokine responses to DNA and DNA viruses. Ref.: Zhang, Z., et al. (2011). Nat. Immunol. 12(10): 959-966.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX41
Applications Western Blot
Classification Monoclonal
Clone 4D8.2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene DDX41, ABS
Gene Accession No. Q9UJV9
Host Species Mouse
Immunogen GST-tagged recombinant fragment corresponding to 115 amino acids from the N-terminal region of human DDX41.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_057306
Test Specificity Clone 4D8.2 detects DDX41 protein in HEPG2 cells by Western blotting and in human ovarian and cervical cancer tissue by immunohisotchemistry.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.