Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX41 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582601
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582601 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582601 Supplier Invitrogen™ Supplier No. PA582601
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEESEPERRR ARTDEVPAGG SRSEAEDEDD EDYVPYVPLR QRRQLLLQKL LQRRRKGAAE EEQQDSGSEP RGDEDDIPLG PQSNVSLLDQ HQHL Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 95%.

Probable ATP-dependent RNA helicase DDX41 is a DEAD box protein characterized by the conserved motif Asp-Glu-Ala-Asp. It is required during post-transcriptional gene expression and may be involved in pre-mRNA splicing. Mutations affecting the gene can result in myeloproliferative/lymphoproliferative neoplasms, familial (MPLPF).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX41
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DDX41
Gene Accession No. Q91VN6, Q9UJV9
Gene Alias 2900024F02Rik; AA958953; ABS; Abstrakt; AI324246; Ddx41; DEAD (Asp-Glu-Ala-Asp) box polypeptide 41; DEAD box protein 41; DEAD box protein abstrakt homolog; DEAD-box helicase 41; DEAD-box protein abstrakt; MPLPF; probable ATP-dependent RNA helicase DDX41; putative RNA helicase
Gene Symbols DDX41
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DDX41. Recombinant protein control fragment (Product #RP-92086).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 314336, 51428, 72935
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.