Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX42 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP187135
Description
DDX42 Polyclonal specifically detects DDX42 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
DDX42 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500-1:1000 | |
DEAD (Asp-Glu-Ala-Asp) box polypeptide 42, DEAD box protein 42, EC 3.6.1, EC 3.6.4.13, RHELPSF3b DEAD box protein, RNA helicase-like protein, RNA helicase-related protein, RNAHPFLJ43179, SF3b125 DEAD-box protein, SF3b125ATP-dependent RNA helicase DDX42, Splicing factor 3B-associated 125 kDa protein | |
Rabbit | |
Affinity Purified | |
RUO | |
11325 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
DDX42 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:HGETGNRHSDSPRHGDGGRHGDGYRHPESSSRHTDGHRHGENRHGGSAGRHGENRGANDGRNGESRKEAFNRESKMEPKMEPKVDSSKMDKVDSKTDKTADGFAVPEPPKRKKSR | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction