Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX43 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128701C
Description
DDX43 Polyclonal antibody specifically detects DDX43 in Human samples. It is validated for Western BlotSpecifications
DDX43 | |
Polyclonal | |
Western Blot | |
Cancer/testis antigen 13, CT13DEAD-box protein 43, DEAD (Asp-Glu-Ala-Asp) box polypeptide 43, DEAD box protein 43, DKFZp434H2114, EC 3.6.1, EC 3.6.4.13, HAGEDEAD box protein HAGE, Helical antigen, probable ATP-dependent RNA helicase DDX43 | |
The immunogen recognized by this antibody maps to a region between residue 275 and 325 of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 43 using the numbering given in entry NP_061135.1 (GeneID 55510). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
55510 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction