Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX43 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590766
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590766 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590766 Supplier Invitrogen™ Supplier No. PA590766
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TWPHSVHRLA QSYLKEPMIV YVGTLDLVAV SSVKQNIIVT TEEEKWSHMQ TFLQSMSSTD KVIVFVSRKA VADHLSSDLI LGNISVESLH GDREQRDREK ALENFKTGKV RILIATDLAS RGLDVHDVTH VYNFDFPRNI EEYVHRIGRT GRAGRTGVSI TTLTRNDWRV ASELINILER ANQSIPEELV SMAERFKAHQ QKREMERKME RPQGRPKKFH; Positive Samples: Mouse testis, Mouse thymus, Rat testis.

The protein encoded by this gene is an ATP-dependent RNA helicase in the DEAD-box family and displays tumor-specific expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX43
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DDX43
Gene Alias Cancer/testis antigen 13; CT13; DDX43; DEAD (Asp-Glu-Ala-Asp) box polypeptide 43; DEAD box protein 43; DEAD box protein HAGE; DEAD-box helicase 43; DEAD-box protein 43; ENSMUSG00000070291; HAGE; Helical antigen; OTTMUSG00000019690; probable ATP-dependent RNA helicase DDX43
Gene Symbols DDX43
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 429-648 of human DDX43 (NP_0611352).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100048658, 682138
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.