Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DDX43 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550396
Description
The antibody detects endogenous levels of total DDX43 protein.
The protein encoded by this gene is an ATP-dependent RNA helicase in the DEAD-box family and displays tumor-specific expression.
Specifications
DDX43 | |
Polyclonal | |
Unconjugated | |
DDX43 | |
Cancer/testis antigen 13; CT13; DDX43; DEAD (Asp-Glu-Ala-Asp) box polypeptide 43; DEAD box protein 43; DEAD box protein HAGE; DEAD-box helicase 43; DEAD-box protein 43; ENSMUSG00000070291; HAGE; Helical antigen; OTTMUSG00000019690; probable ATP-dependent RNA helicase DDX43 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55510 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2.3 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9NXZ2 | |
DDX43 | |
Fusion protein corresponding to a region derived from internal residues of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 43. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction