Learn More
DDX43 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302328AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene is an ATP-dependent RNA helicase in the DEAD-box family and displays tumor-specific expression.Specifications
DDX43 | |
Polyclonal | |
Unconjugated | |
DDX43 | |
CT13, HAGE, DEAD box protein 43, DEAD box protein HAGE, DEAD-box protein 43, cancer/testis antigen 13, helical antigen, probable ATP-dependent RNA helicase DDX43 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55510 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9NXZ2 | |
DDX43 | |
Between 275 and 325 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.