Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX46 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10057489V
Description
DDX46 Polyclonal antibody specifically detects DDX46 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
DDX46 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
DEAD (Asp-Glu-Ala-Asp) box polypeptide 46, DEAD box protein 46, EC 3.6.1, EC 3.6.4.13, FLJ25329, KIAA0801probable ATP-dependent RNA helicase DDX46, MGC9936, Prp5, PRP5 homolog, Prp5-like DEAD-box protein, PRPF5, RNA helicase | |
The immunogen recognized by this antibody maps to a region between residue 300 and 350 of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 46 using the numbering given in entry NP_055644.2 (GeneID 9879). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
9879 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction