Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX46 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10057490
Description
DDX46 Polyclonal specifically detects DDX46 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
DDX46 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
DEAD (Asp-Glu-Ala-Asp) box polypeptide 46, DEAD box protein 46, EC 3.6.1, EC 3.6.4.13, FLJ25329, KIAA0801probable ATP-dependent RNA helicase DDX46, MGC9936, Prp5, PRP5 homolog, Prp5-like DEAD-box protein, PRPF5, RNA helicase | |
Rabbit | |
Affinity Purified | |
RUO | |
9879 | |
Store at 4C. Do not freeze. |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunohistochemistry 1:100 - 1:500, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:100 - 1:500 | |
Q7L014 | |
DDX46 | |
The immunogen recognized by this antibody maps to a region between residue 981 and the C-terminus (residue 1031) of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 46 using the numbering given in entry NP_055644.2 (GeneID 9879). | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction