Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX46 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557713
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557713 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557713 Supplier Invitrogen™ Supplier No. PA557713
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KEGNEMEGEE LDPLDAYMEE VKEEVKKFNM RSVKGGGGNE KKSGPTVTKV VTVVTTKKAV VDSDKKKGEL MENDQDAM Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 96%.

This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The protein encoded by this gene is a component of the 17S U2 snRNP complex; it plays an important role in pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX46
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DDX46
Gene Accession No. Q7L014
Gene Alias 2200005K02Rik; 8430438J23Rik; AI325430; AI957095; DDX46; DEAD (Asp-Glu-Ala-Asp) box polypeptide 46; DEAD box polypeptide 46; DEAD box protein 46; DEAD-box helicase 46; Hel117; Helicase of 117.4 kDa; Kiaa0801; mKIAA0801; Probable ATP-dependent RNA helicase DDX46; Prp5; PRP5 homolog; Prp5-like DEAD-box protein; PRPF5; RNA helicase
Gene Symbols DDX46
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DDX46. Recombinant protein control fragment (Product #RP-96886).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9879
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.