Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX47 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587840
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587840 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587840 Supplier Invitrogen™ Supplier No. PA587840
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TEVDKILKVI PRDRKTFLFS ATMTKKVQKL QRAALKNPVK CAVSSKYQTV EKLQQYYIFI PSKFKDTYLV YILNELAGNS FMIFCSTCNN TQRTALLLRN LGFTAIPLHG QMSQSKRLGS LNKFKAKARS ILLATDVASR GLDIPHVDVV VNFDIPTHSK DYIHRVGRTA RAGRSGKAIT FVTQYDVELF QRIEHLIGKK LPGFPTQDDE VMMLTERVAE AQRFARMELR EHGEKKKRSR EDAGDNDDTE GAIGVRNKVA GGKMKKRKGR; Positive Samples: U-87MG, SKOV3, LO2, HeLa, Mouse testis, Mouse lung, Rat testis; Cellular Location: Nucleus, nucleolus.

This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The protein encoded by this gene can shuttle between the nucleus and the cytoplasm, and has an RNA-independent ATPase activity. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX47
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DDX47
Gene Accession No. Q9CWX9, Q9H0S4
Gene Alias 4930588A18Rik; C77285; DDX47; DEAD (Asp-Glu-Ala-Asp) box polypeptide 47; DEAD box polypeptide 47; DEAD box protein 47; DEAD-box helicase 47; E4-DBP; E4-DEAD box protein; HQ0256; MSTP162; Probable ATP-dependent RNA helicase DDX47; RRP3
Gene Symbols DDX47
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 186-455 of human DDX47 (NP_0574392).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 297685, 51202, 67755
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.