Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DDX49 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531274
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (91%), Rat (90%), Chicken (83%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The function of DDX49 has not yet been determined.
Specifications
| DDX49 | |
| Polyclonal | |
| Unconjugated | |
| DDX49 | |
| DDX49; DEAD (Asp-Glu-Ala-Asp) box polypeptide 49; DEAD box protein 49; DEAD-box helicase 49; probable ATP-dependent RNA helicase DDX49; R27090_2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54555 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.77 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9Y6V7 | |
| DDX49 | |
| Recombinant protein encompassing a sequence within the center region of human DDX49. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction