Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX49 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559765
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559765 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559765 Supplier Invitrogen™ Supplier No. PA559765
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TNQPFFWEAQ APVSTVEQLD QRYLLVPEKV KDAYLVHLIQ RFQDEHEDWS IIIFTNTCKT CQILCMMLRK FSFPTVALHS MMKQKE Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.

The function of DDX49 has not yet been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX49
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DDX49
Gene Accession No. Q9Y6V7
Gene Alias DDX49; DEAD (Asp-Glu-Ala-Asp) box polypeptide 49; DEAD box protein 49; DEAD-box helicase 49; probable ATP-dependent RNA helicase DDX49; R27090_2
Gene Symbols DDX49
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DDX49. Recombinant protein control fragment (Product #RP-99000).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54555
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.