Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
Supplier: Novus Biologicals NB200351
Description
DDX5 Polyclonal specifically detects DDX5 in Human, Mouse, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
DDX5 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
B-cell CLL/lymphoma 7 protein family member C, B-cell CLL/lymphoma 7C | |
Rabbit | |
69 kDa | |
0.1 mL | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Gorilla, Chimpanzee, White-tufted-earmarmoset, Chinese hamster, Naked mole rat, African elephant, and Bonnet macaque. | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
Western Blot 1:2500-1:10000, Immunohistochemistry 1:200 - 1:1000, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1 ug/ml | |
P17844 | |
DDX5 | |
The immunogen recognized by this antibody maps to a region between residue 575 and the C-terminus (residue 614) of human DEAD (Asp-Glu-Ala-Asp) Polypeptide 5 using the numbering given in entry NP_004387.1 (GeneID 1655). | |
Affinity Purified | |
RUO | |
1655 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction