Learn More
DDX5 Mouse anti-Human, Unconjugated, Clone: 2D3D2, Proteintech
Shop All Proteintech Group Inc Products

Description
The DDX5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DDX5. This antibody recognizes human, mouse, rat antigen. The DDX5 antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.
Specifications
Specifications
| Antigen | DDX5 |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Clone | 2D3D2 |
| Concentration | 1000 μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 150 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 1655 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.