Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX50 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen DDX50
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP157289
SDP
View Documents
Novus Biologicals
NBP157289
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

DDX50 Polyclonal specifically detects DDX50 in Human samples. It is validated for Western Blot.
Specifications

Specifications

DDX50
Polyclonal
Purified
RUO
ATP-dependent RNA helicase DDX50, DEAD (Asp-Glu-Ala-Asp) box polypeptide 50, DEAD box protein 50, EC 3.6.1, EC 3.6.4.13, GU2, GUB, gu-beta, MGC3199, Nucleolar protein Gu2, RH-II/GuB, RNA helicase II/Gu beta
DDX50
IgG
Protein A purified
Western Blot
Unconjugated
Rabbit
Q9BQ39
79009
Synthetic peptides corresponding to DDX50 (DEAD (Asp-Glu-Ala-Asp) box polypeptide 50) The peptide sequence was selected from the N terminal of DDX50. Peptide sequence EESESQKKERQKSDRRKSRHHYDSDEKSETRENGVTDDLDAPKAKKSKMK.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.