Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX50 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297538PE
Description
DDX50 Polyclonal antibody specifically detects DDX50 in Human samples. It is validated for Western BlotSpecifications
DDX50 | |
Polyclonal | |
Western Blot | |
ATP-dependent RNA helicase DDX50, DEAD (Asp-Glu-Ala-Asp) box polypeptide 50, DEAD box protein 50, EC 3.6.1, EC 3.6.4.13, GU2, GUB, gu-beta, MGC3199, Nucleolar protein Gu2, RH-II/GuB, RNA helicase II/Gu beta | |
Produced in rabbits immunized with E. coli-derived Human DDX50 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
79009 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction