Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX50 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597136
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597136 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597136 Supplier Invitrogen™ Supplier No. PA597136
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPGKLLWGDI MELEAPLEES ESQKKERQKS DRRKSRHHYD SDEKSETREN GVTDDLDAPK AKKSKMKEKL NGDTEEGFNR LSDEFSKSHK SRRKDLPNGD IDEYEKKSKR VSSLDTSTHK SSDNKLEETL TREQK; Positive Samples: A-431, Jurkat, HepG2; Cellular Location: Nucleus, nucleolus.

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. DDX50 is a DEAD box enzyme that may be involved in ribosomal RNA synthesis or processing. DDX50 and DDX21, also called RH-II/GuA, have similar genomic structures and are in tandem orientation on chromosome 10, suggesting that the two genes arose by gene duplication in evolution. DDX50 gene has pseudogenes on chromosomes 2, 3 and 4. Alternative splicing of this gene generates multiple transcript variants, but the full length nature of all the other variants but one has not been defined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX50
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DDX50
Gene Accession No. Q99MJ9, Q9BQ39
Gene Alias 4933429B04Rik; 8430408E17Rik; ATP-dependent RNA helicase DDX50; ATP-dependent RNA helicase DDX50-like protein; Ddx50; DEAD (Asp-Glu-Ala-Asp) box polypeptide 50; DEAD box protein 50; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 50; DEAD-box helicase 50; DExD-box helicase 50; GU2; GUB; Gubeta; gu-beta; malignant cell derived RNA helicase; mcdrh; Nucleolar protein Gu2; RH-II; RH-II/GuB; RH-II/Gubeta; RNA helicase II/Gu beta; Unknown (protein for IMAGE:8120580)
Gene Symbols DDX50
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-135 of human DDX50 (NP_0769501).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361848, 79009, 94213
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.