Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DDX50 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598827
Description
DDX50 Polyclonal Antibody for Western Blot, IHC (P), ELISA
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. DDX50 is a DEAD box enzyme that may be involved in ribosomal RNA synthesis or processing. DDX50 and DDX21, also called RH-II/GuA, have similar genomic structures and are in tandem orientation on chromosome 10, suggesting that the two genes arose by gene duplication in evolution. DDX50 gene has pseudogenes on chromosomes 2, 3 and 4. Alternative splicing of this gene generates multiple transcript variants, but the full length nature of all the other variants but one has not been defined.
Specifications
DDX50 | |
Polyclonal | |
Unconjugated | |
DDX50 | |
4933429B04Rik; 8430408E17Rik; ATP-dependent RNA helicase DDX50; ATP-dependent RNA helicase DDX50-like protein; Ddx50; DEAD (Asp-Glu-Ala-Asp) box polypeptide 50; DEAD box protein 50; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 50; DEAD-box helicase 50; DExD-box helicase 50; GU2; GUB; Gubeta; gu-beta; malignant cell derived RNA helicase; mcdrh; Nucleolar protein Gu2; RH-II; RH-II/GuB; RH-II/Gubeta; RNA helicase II/Gu beta; Unknown (protein for IMAGE:8120580) | |
Rabbit | |
Protein G | |
RUO | |
79009 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
4 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9BQ39 | |
DDX50 | |
Recombinant Human ATP-dependent RNA helicase DDX50 protein (1-210AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction