Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX51 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10057526
Description
DDX51 Polyclonal specifically detects DDX51 in Human samples. It is validated for Immunoprecipitation.Specifications
DDX51 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
ATP-dependent RNA helicase DDX51, DEAD (Asp-Glu-Ala-Asp) box polypeptide 51, DEAD box protein 51, DKFZp686N2081, EC 3.6.1, EC 3.6.4.13, MGC42193 | |
Rabbit | |
Affinity Purified | |
RUO | |
317781 | |
Store at 4C. Do not freeze. |
Immunoprecipitation | |
0.1 mg/mL | |
Immunoprecipitation 2 - 5 ug/mg lysate | |
NP_778236.1 | |
DDX51 | |
The immunogen recognized by this antibody maps to a region between residue 75 and 125 of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 51 using the numbering given in entry NP_778236.1 (GeneID 317781). | |
100 μL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction