Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX51 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590569
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590569 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590569 Supplier Invitrogen™ Supplier No. PA590569
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FLIIDEADRM IDSMHQSWLP RVVAAAFQSE DPADPCALLQ RRQAQAVTAA STCCPQMPLQ KLLFSATLTQ NPEKLQQLGL HQPRLFSTGL AHRGLEDTDG DGDSGKYAFP VGLTHHYVPC SLSSKPLVVL HLVLEMGFSR VLCFTNSREN SHRLFLLVQA FGGVDVAEFS SRYGPGQRRM ILKQFEQGKI QLLISTDATA RGIDVQGVEL VVNYDAPQYL RTYVHRVGRT ARAGKTGQAF TLLLKVQERR FLRMLTEAGA PELQRHELSS KLLQPLVPRY EEALSQLEES VKEERKQRAA; Positive Samples: HT-29, Jurkat, Mouse brain, Mouse spleen, Rat testis; Cellular Location: Nucleus, nucleolus.

ATP-binding RNA helicase involved in the biogenesis of 60S ribosomal subunits. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX51
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DDX51
Gene Accession No. Q6P9R1, Q8N8A6
Gene Alias 2310061O04Rik; AA537002; ATP-dependent RNA helicase DDX51; C81583; Ddx51; DEAD (Asp-Glu-Ala-Asp) box polypeptide 51; DEAD box protein 51; DEAD-box helicase 51; R75505
Gene Symbols DDX51
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 367-666 of human DDX51 (NP_7782362).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 304570, 317781, 69663
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.