Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX51 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101448
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101448 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101448 Supplier Invitrogen™ Supplier No. PA5101448
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DDX51.

ATP-binding RNA helicase involved in the biogenesis of 60S ribosomal subunits. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX51
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DDX51
Gene Accession No. Q8N8A6
Gene Alias 2310061O04Rik; AA537002; ATP-dependent RNA helicase DDX51; C81583; Ddx51; DEAD (Asp-Glu-Ala-Asp) box polypeptide 51; DEAD box protein 51; DEAD-box helicase 51; R75505
Gene Symbols DDX51
Host Species Rabbit
Immunogen A synthesized peptide derived from human DDX51(Accession Q8N8A6), corresponding to amino acid residues H632-A666.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 317781
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.