Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX51 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA558817

Catalog No. PIPA558817


Only null left
Add to cart

Description

Description

Immunogen sequence: GFGKRKAPKV QPFLPRWLAE PNCVRRNVTE DLVPIEDIPD VHPDLQKQLR AHGISSYFPV QAAVIPALLE SAACGFLVGR GGYRPSDLCV SA Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 82%.

ATP-binding RNA helicase involved in the biogenesis of 60S ribosomal subunits. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DDX51
Polyclonal
Unconjugated
DDX51
2310061O04Rik; AA537002; ATP-dependent RNA helicase DDX51; C81583; Ddx51; DEAD (Asp-Glu-Ala-Asp) box polypeptide 51; DEAD box protein 51; DEAD-box helicase 51; R75505
Rabbit
Antigen affinity chromatography
RUO
317781
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8N8A6
DDX51
Recombinant protein corresponding to Human DDX51. Recombinant protein control fragment (Product #RP-97872).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.