Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX52 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171810AF594
Description
DDX52 Polyclonal antibody specifically detects DDX52 in Human samples. It is validated for Western BlotSpecifications
DDX52 | |
Polyclonal | |
Western Blot | |
ATP-dependent RNA helicase ROK1-like, DEAD (Asp-Glu-Ala-Asp) box polypeptide 52, DEAD box protein 52, EC 3.6.1, HUSSY19, probable ATP-dependent RNA helicase DDX52, ROK1EC 3.6.4.13 | |
The immunogen for this product maps to a region between residue 499 and 549 of human DEAD (Asp-Glu-Ala-Asp) Box Polypeptide 52 using the numbering given in entry NP_008941.2 (GeneID 11056). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
11056 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction